Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sos
- Entrez Gene ID
-
34790
- Gene Name
- Son of sevenless
- Gene Aliases
-
2.3, 34Ea, BG:DS00941.4, CG7793, Dmel\CG7793, Dmel_CG7793, E(sev)2A, E(var)189, EC2-5, EK2-8, EY2-3, SOS, Sos1, Su(tor)2-2, br24, br25, dSos, dme-SOS, l(2)34Ea, l(2)Sos, l(2)br24, l(2)br25, l(2)k05224, l(2)k06321, sos
- Chromosome Location
-
Chr.2L: 13813816 - 13819824 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001965
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057249.6 |
NP_476597.2 |
6 - 7 |
5143 |
111 |
View Full Product Details