Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mle
- Entrez Gene ID
-
35523
- Gene Name
- maleless
- Gene Aliases
-
CG11680, Dmel\CG11680, Dmel_CG11680, MLE, Mle, dDHX9, mak, mll, mls, nap, nap[ts]
- Chromosome Location
-
Chr.2R: 5974857 - 5980778 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002774
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057293.4 |
NP_476641.1 |
2 - 3 |
675 |
88 |
GenBank mRNA |
BT010267.1 |
- |
2 - 3 |
671 |
88 |
View Full Product Details