Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rk
- Entrez Gene ID
-
34819
- Gene Name
- rickets
- Gene Aliases
-
BG:DS00180.13, CG8930, CT25644, DLGR-2, DLGR2, DLgr2, Dmel\CG8930, Dmel_CG8930, LGR2, Lgr2, Rk, dlgr2, dlgr2/rk, lgr2, rk/CG8930
- Chromosome Location
-
Chr.2L: 13982743 - 14001248 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003255
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057354.3 |
NP_476702.1 |
4 - 5 |
1391 |
62 |
View Full Product Details