Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- shg
- Entrez Gene ID
-
37386
- Gene Name
- shotgun
- Gene Aliases
-
CADH, CG3722, CT12481, Cad, CadE, Cadh, D E-cad, D-CAD2, D-cad, DCAD, DCAD2, DCad, DCad2, DE, DE Cad, DE Cadh, DE-CAD, DE-CAD2, DE-Cad, DE-Cad2, DE-Cadh, DE-Cadherin, DE-cad, DE-cadh, DE-cadherin, DEC, DECAD, DECad, DECadh, DE[cyto], DEcad, De-Cad, Dmel\CG3722, Dmel_CG3722, E-CAD, E-Ca, E-Cad, E-Cadherin, E-cad, E-cadherin, ECAD, ECad, ECad2, ECadh, Ecad, N-cad, Shg, cad, cadh, com5, dEcad, de-cad, e-cad, ecad, gp150, l(2)10469, l(2)k03401, l(2)k10220, shg/DE-Cad
- Chromosome Location
-
Chr.2R: 21049654 - 21057197 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003391
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057374.3 |
NP_476722.1 |
1 - 2 |
1352 |
69 |
View Full Product Details