Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tra2
- Entrez Gene ID
-
36619
- Gene Name
- transformer 2
- Gene Aliases
-
CG10128, Dmel\CG10128, Dmel_CG10128, TRA 2, TRA-2, TRA2, Tra-2, Tra2, tra-2
- Chromosome Location
-
Chr.2R: 14602004 - 14604337 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003742
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057415.3 |
NP_476763.1 |
4 - 5 |
565 |
71 |
GenBank mRNA |
M76381.1 |
- |
4 - 5 |
585 |
71 |
View Full Product Details