Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Uro
- Entrez Gene ID
-
34060
- Gene Name
- Urate oxidase
- Gene Aliases
-
CG7171, Dm UO, Dmel\CG7171, Dmel_CG7171, OU, UO, UOX, Uo, anon-WO0140519.210, uro
- Chromosome Location
-
Chr.2L: 7780085 - 7781415 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003961
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057431.3 |
NP_476779.1 |
1 - 2 |
606 |
61 |
View Full Product Details