Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vm26Aa
Entrez Gene ID
33829
Gene Name
Vitelline membrane 26Aa
Gene Aliases
CG9048, Dmel\CG9048, Dmel_CG9048, TU-2, VM26A.1, VM26Aa, Vm26A, Vm26A.1, Vm26A1, sV17
Chromosome Location
Chr.2L: 5959596 - 5960340 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003979
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273186.1 NP_001260115.1 1 - 1 544 79
NM_057435.3 NP_476783.1 1 - 1 544 79
GenBank mRNA BT025155.1 - 1 - 1 522 79

View Full Product Details