Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPO1
- Entrez Gene ID
-
37044
- Gene Name
- Prophenoloxidase 1
- Gene Aliases
-
A1, A[[1]], Bc, CG42639, CG5779, DmePPOA1, Dmel\CG42639, Dmel_CG42639, Dox-A1, DoxA1, Mox, PO, PO A1, PO54, PPO, PPO-1, PROPO-A1, Phox, ProPO, ProPO A1, ProPO54, ProPo, Propo, bc, i12, pro-A, pro-PO, pro-POA[[1]], proPO, proPO A[[1]], proPO-A1, proPO54
- Chromosome Location
-
Chr.2R: 17887220 - 17889973 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0283437
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057464.4 |
NP_476812.1 |
4 - 5 |
1700 |
69 |
GenBank mRNA |
AY060617.1 |
- |
4 - 5 |
1654 |
69 |
View Full Product Details