Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mdr65
Entrez Gene ID
38726
Gene Name
Multi drug resistance 65
Gene Aliases
CG10181, Dmel\CG10181, Dmel_CG10181, MDR65, Mdr65A, P-glycoprotein, P-gp, mdr65
Chromosome Location
Chr.3L: 6240500 - 6245746 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004513
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057483.4 NP_476831.1 11 - 12 3480 81

View Full Product Details