Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mdr65
- Entrez Gene ID
-
38726
- Gene Name
- Multi drug resistance 65
- Gene Aliases
-
CG10181, Dmel\CG10181, Dmel_CG10181, MDR65, Mdr65A, P-glycoprotein, P-gp, mdr65
- Chromosome Location
-
Chr.3L: 6240500 - 6245746 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004513
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057483.4 |
NP_476831.1 |
3 - 4 |
684 |
134 |
View Full Product Details