Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- inaC
- Entrez Gene ID
-
36897
- Gene Name
- inactivation no afterpotential C
- Gene Aliases
-
CG6518, Dmel\CG6518, Dmel_CG6518, Dpkc2, INAC, InaC, PK-C, PKC, PKC 53E, PKC2, PKCi, PKCinac, Pkc2, dPKC53E, dPKC53E(ey), ePKC, eye PKC, eye-PKC, eyePKC, inac, pkc-2
- Chromosome Location
-
Chr.2R: 16898535 - 16901575 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004784
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057515.3 |
NP_476863.1 |
4 - 5 |
743 |
71 |
GenBank mRNA |
BT004495.1 |
- |
4 - 5 |
701 |
71 |
View Full Product Details