Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
U2af50
Entrez Gene ID
32602
Gene Name
U2 small nuclear riboprotein auxiliary factor 50
Gene Aliases
9-21, CG9998, DU2AF50, DmU2AF[50], Dmel\CG9998, Dmel_CG9998, U2AF, U2AF 50, U2AF-50, U2AF50, U2af, dU2AF50, dU2AF[50], dUTAf[50], l(1)14Ca, l(1)14Cc, l(1)9-21
Chromosome Location
Chr.X: 16356696 - 16359039 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005411
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258779.1 NP_001245708.1 4 - 5 823 90
NM_001258780.1 NP_001245709.1 4 - 5 850 90
NM_001258781.2 NP_001245710.1 4 - 5 952 90
NM_057543.4 NP_476891.1 4 - 5 1006 90
GenBank mRNA AY069320.1 - 4 - 5 916 90
L23404.1 - 4 - 5 917 90

View Full Product Details