Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pelo
- Entrez Gene ID
-
34286
- Gene Name
- pelota
- Gene Aliases
-
CG3959, DmPelota, Dmel\CG3959, Dmel_CG3959, PELO, Pelo, Pelo Hbs1, ms(2)01559
- Chromosome Location
-
Chr.2L: 9700810 - 9708883 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011207
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057634.5 |
NP_476982.1 |
4 - 5 |
1303 |
66 |
GenBank mRNA |
AY058644.1 |
- |
4 - 5 |
1320 |
66 |
View Full Product Details