Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Elk
- Entrez Gene ID
-
37047
- Gene Name
- Eag-like K[+] channel
- Gene Aliases
-
CG5076, Dmel\CG5076, Dmel_CG5076, dmELK, elk
- Chromosome Location
-
Chr.2R: 17873939 - 17926695 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011589
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057661.3 |
NP_477009.1 |
10 - 11 |
1508 |
83 |
View Full Product Details