Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ine
Entrez Gene ID
33659
Gene Name
inebriated
Gene Aliases
CG15444, CG15444d, DMINE, Dmel\CG15444, Dmel_CG15444, ine/rosA, rosA
Chromosome Location
Chr.2L: 4468000 - 4477022 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011603
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169400.2 NP_001162871.1 5 - 6 1749 81
NM_057664.6 NP_477012.1 5 - 6 1947 81
NM_057665.5 NP_477013.1 3 - 4 555 81
NM_175958.3 NP_787972.1 5 - 6 1735 81
NM_175959.3 NP_787973.1 5 - 6 1771 81
GenBank mRNA AY128432.1 - 5 - 6 1751 81
BT009955.1 - 5 - 6 1773 81
BT099793.1 - 3 - 4 538 81

View Full Product Details