Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ine
Entrez Gene ID
33659
Gene Name
inebriated
Gene Aliases
CG15444, CG15444d, DMINE, Dmel\CG15444, Dmel_CG15444, ine/rosA, rosA
Chromosome Location
Chr.2L: 4468000 - 4477022 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011603
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169400.2 NP_001162871.1 6 - 7 1885 60
NM_057664.6 NP_477012.1 6 - 7 2083 60
NM_057665.5 NP_477013.1 4 - 5 691 60
NM_175958.3 NP_787972.1 6 - 7 1871 60
NM_175959.3 NP_787973.1 6 - 7 1907 60
GenBank mRNA AY128432.1 - 6 - 7 1887 60
BT009955.1 - 6 - 7 1909 60
BT099793.1 - 4 - 5 674 60
U66460.1 - 4 - 5 650 60

View Full Product Details