Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wee1
Entrez Gene ID
33965
Gene Name
Wee1 kinase
Gene Aliases
CG4488, Dmel\CG4488, Dmel_CG4488, Dwee, Dwee1, WEE1, Wee, Wee 1, dWee1, dwee1, wee, wee1
Chromosome Location
Chr.2L: 6911090 - 6914138 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011737
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273238.1 NP_001260167.1 4 - 5 1887 78
NM_057687.4 NP_477035.1 4 - 5 1887 78

View Full Product Details