Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Arp53D
- Entrez Gene ID
-
36879
- Gene Name
- Actin-related protein 53D
- Gene Aliases
-
ARP53D, Actr53D, ArpG, Arp_G, CG5409, Dmel\CG5409, Dmel_CG5409, arp5, arp53D
- Chromosome Location
-
Chr.2R: 16774246 - 16775676 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011743
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057689.4 |
NP_477037.2 |
2 - 2 |
1195 |
62 |
View Full Product Details