Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dp
- Entrez Gene ID
-
36461
- Gene Name
- DP transcription factor
- Gene Aliases
-
49Fk, CG4654, DDP1, DP, DP1, DmDP, Dmel\CG4654, Dmel_CG4654, Dp1, TFPD2, dDP, dDP1, dDp, dp, l(2)49Fk, l(2)vr10, vr10
- Chromosome Location
-
Chr.2R: 13222433 - 13228202 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011763
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057691.4 |
NP_477039.1 |
1 - 2 |
681 |
64 |
GenBank mRNA |
AY069526.1 |
- |
1 - 2 |
630 |
64 |
View Full Product Details