Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pen
- Entrez Gene ID
-
34338
- Gene Name
- Pendulin
- Gene Aliases
-
2.1, CG4799, DPend, Dalpha2, Dimp-alpha2, Dmel\CG4799, Dmel_CG4799, IMPalpha2, Imp-alpha2, Impalpha2, Kap alpha2, Kap-alpha2, Kpna2, OHO31, Oho31, Rch1, alpha2, alpha2A-Kap, alphaKap2, anon-WO0140519.258, bs29g06.y1, imp alpha2, imp-alpha2, impalpha2, importin alpha2/pendulin, l(2)144/1, l(2)k06324, l(2)k14401, oho-31, oho31, pen, pen-2
- Chromosome Location
-
Chr.2L: 10056900 - 10060095 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0267727
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057693.5 |
NP_477041.1 |
3 - 4 |
758 |
86 |
GenBank mRNA |
BT003258.1 |
- |
3 - 4 |
668 |
86 |
View Full Product Details