Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rh5
Entrez Gene ID
34615
Gene Name
Rhodopsin 5
Gene Aliases
CG5279, DMELRH5, Dm Rh5, Dmel\CG5279, Dmel_CG5279, RH5, Rh, rh5
Chromosome Location
Chr.2L: 12008153 - 12009832 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014019
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298930.1 NP_001285859.1 2 - 3 882 82
NM_057748.5 NP_477096.1 2 - 3 657 82
GenBank mRNA AY069237.1 - 2 - 3 643 82

View Full Product Details