Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- csw
- Entrez Gene ID
-
45278
- Gene Name
- corkscrew
- Gene Aliases
-
19-106, CG3954, CSW, Csw, Csw/Shp2, Dmel\CG3954, Dmel_CG3954, E(sev)1A, EG:BACN25G24.2, SHP-2, Shp-2, Shp2/csw, anon-WO03040301.207, anon-WO03040301.209, anon-WO03040301.219, csw/SHP-2, l(1)2Db, l(1)2Dd, l(1)G0170, l(1)GA114, l(1)csw
- Chromosome Location
-
Chr.X: 2094234 - 2115701 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000382
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057783.3 |
NP_477131.1 |
1 - 2 |
1483 |
99 |
GenBank mRNA |
BT149891.1 |
- |
1 - 2 |
1287 |
99 |
View Full Product Details