Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pi3K59F
- Entrez Gene ID
-
37733
- Gene Name
- Phosphatidylinositol 3-kinase 59F
- Gene Aliases
-
CG5373, DmVps34, Dmel\CG5373, Dmel_CG5373, PI(3)K, PI3K, PI3K-59F, PI3K59F, PI3K_59F, PIK359F, PI[[3]]K, Pi3K_59F, Pi3k59F, VPS34, Vps34, dPI3K, dVps34, vps34
- Chromosome Location
-
Chr.2R: 23560492 - 23564045 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015277
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057785.5 |
NP_477133.1 |
1 - 2 |
155 |
58 |
GenBank mRNA |
AY058362.1 |
- |
1 - 2 |
132 |
58 |
View Full Product Details