Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lic
Entrez Gene ID
32257
Gene Name
licorne
Gene Aliases
CG12244, D-MKK3, DMEK3, DMKK3, DMKK3/lic, DmMKK3, Dmel\CG12244, Dmel_CG12244, MEK3, MEK3/MKK3, MKK3, Mek3, Mkk3, Mpk3, dMKK3, l(1)G0252, p38, p38MAPKK
Chromosome Location
Chr.X: 13091145 - 13093986 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261524
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057814.4 NP_477162.1 4 - 5 1120 105
GenBank mRNA AF035550.1 - 4 - 5 1042 105
AF041136.1 - 4 - 5 1107 105
AY052103.1 - 4 - 5 1102 105

View Full Product Details