Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Reg-5
- Entrez Gene ID
-
37968
- Gene Name
- Rhythmically expressed gene 5
- Gene Aliases
-
CG2928, DREG-5, Dmel\CG2928, Dmel_CG2928, Dreg, Dreg-5, Dreg5, dreg-5, reg-5
- Chromosome Location
-
Chr.2R: 24788008 - 24793410 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015801
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057825.5 |
NP_477173.1 |
2 - 3 |
816 |
57 |
GenBank mRNA |
AY071459.1 |
- |
2 - 3 |
814 |
57 |
View Full Product Details