Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ric
Entrez Gene ID
36776
Gene Name
Ras-related protein interacting with calmodulin
Gene Aliases
17137290, CG8418, D-Ric, Dmel\CG8418, Dmel_CG8418, Q7JMZ0, RIC, Roc
Chromosome Location
Chr.2R: 16107440 - 16110017 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265605
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169694.2 NP_001163165.1 2 - 3 444 61
NM_057863.4 NP_477211.1 2 - 3 444 61
GenBank mRNA Y07564.1 - 2 - 3 395 61

View Full Product Details