Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pkg21D
- Entrez Gene ID
-
33253
- Gene Name
- Protein kinase, cGMP-dependent at 21D
- Gene Aliases
-
CG3324, DG1, Dg1, Dmel\CG3324, Dmel_CG3324, PKG, PKG 21D, PKG-21D, PKG1, PKG21D, Pkg1, cGMP, dg1, pkg21D
- Chromosome Location
-
Chr.2L: 786153 - 790798 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000442
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057865.4 |
NP_477213.1 |
1 - 2 |
309 |
67 |
GenBank mRNA |
AY058288.1 |
- |
1 - 2 |
292 |
67 |
View Full Product Details