Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SamDC
- Entrez Gene ID
-
34396
- Gene Name
- S-adenosylmethionine decarboxylase
- Gene Aliases
-
CG5029, Dmel\CG5029, Dmel_CG5029, l(2)31Dc, l(2)31Dd, l(2)31De
- Chromosome Location
-
Chr.2L: 10339457 - 10341503 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0019932
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057875.5 |
NP_477223.2 |
6 - 7 |
951 |
80 |
GenBank mRNA |
BT021946.1 |
- |
6 - 7 |
937 |
80 |
View Full Product Details