Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ttv
Entrez Gene ID
36614
Gene Name
tout-velu
Gene Aliases
CG10117, DEXT1, Dmel\CG10117, Dmel_CG10117, Ext1, P1.15, Ttv, dExt1, l(2)00681, l(2)05282, l(2)SH0631, l(2)SH2 0631, l(2)k00115, l(2)k03617
Chromosome Location
Chr.2R: 14526267 - 14587917 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265974
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274042.1 NP_001260971.1 4 - 5 2081 59
NM_001369957.1 NP_001356963.1 4 - 5 1772 59
NM_057883.3 NP_477231.1 4 - 5 1772 59
GenBank mRNA AB221351.1 - 4 - 5 1418 59
BT021403.1 - 4 - 5 1777 59

View Full Product Details