Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk
Entrez Gene ID
34843
Gene Name
pickpocket
Gene Aliases
BG:DS06238.1, CG3478, Dmel\CG3478, Dmel_CG3478, PPK, PPK1, dmdNaC1, dmdNaCl, mdNaC1, pkt, ppk1
Chromosome Location
Chr.2L: 14378218 - 14381172 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020258
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057884.5 NP_477232.1 1 - 2 302 66
GenBank mRNA AF043263.1 - 1 - 2 264 66
BT044099.1 - 1 - 2 301 66

View Full Product Details