Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sdk
Entrez Gene ID
31017
Gene Name
sidekick
Gene Aliases
BcDNA:LD22322, BcDNA:RE63453, CG5227, CT16627, Dmel\CG5227, Dmel_CG5227, EG:BACR19J1.1
Chromosome Location
Chr.X: 686834 - 749496 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0021764
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169159.2 NP_001162630.1 9 - 10 3127 80
NM_001169160.2 NP_001162631.1 10 - 11 2495 80
NM_001297827.1 NP_001284756.1 11 - 12 2752 80
NM_001297828.1 NP_001284757.1 10 - 11 2617 80
NM_001297829.1 NP_001284758.1 9 - 10 2461 80
NM_057941.5 NP_477289.5 10 - 11 2564 80
NM_057942.5 NP_477290.6 10 - 11 2681 80
NM_134314.4 NP_599141.5 10 - 11 2629 80
NM_134315.4 NP_599142.5 10 - 11 2681 80

View Full Product Details