Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cht2
Entrez Gene ID
38223
Gene Name
Chitinase 2
Gene Aliases
CG2054, CHT2, DmCHT2, DmCht2, Dmel\CG2054, Dmel_CG2054, cht2
Chromosome Location
Chr.3L: 1752328 - 1755934 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022702
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259621.2 NP_001246550.1 3 - 4 1709 88
NM_001274353.1 NP_001261282.1 4 - 5 1145 88
NM_057950.4 NP_477298.2 4 - 5 879 88
GenBank mRNA AY058589.1 - 4 - 5 836 88

View Full Product Details