Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HLH54F
- Entrez Gene ID
-
37027
- Gene Name
- CG5005-PA
- Gene Aliases
-
CG5005, Dmel\CG5005, Dmel_CG5005, HLH102C, HLH54, HLH54F-Dm, bHLH54F, bHLHa9
- Chromosome Location
-
Chr.2R: 17757405 - 17760517 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0022740
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057954.3 |
NP_477302.1 |
2 - 3 |
602 |
77 |
GenBank mRNA |
BT028823.2 |
- |
2 - 3 |
448 |
77 |
View Full Product Details