Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hs2st
- Entrez Gene ID
-
44433
- Gene Name
- Heparan sulfate 2-O-sulfotransferase
- Gene Aliases
-
CG10234, DmHs2st, Dmel\CG10234, Dmel_CG10234, HS2ST, Hs2st-2, dHS2ST, dmHS-2OST, dmHs2st
- Chromosome Location
-
Chr.2L: 19442833 - 19444750 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024230
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057991.5 |
NP_477339.1 |
3 - 4 |
717 |
66 |
GenBank mRNA |
AY058422.1 |
- |
3 - 4 |
728 |
66 |
View Full Product Details