Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cv-2
- Entrez Gene ID
-
45280
- Gene Name
- crossveinless 2
- Gene Aliases
-
BMPER, CG15671, CT35855, CV2, Cv 2, Cv-2, Cv2, Dmel\CG15671, Dmel_CG15671, cv 2
- Chromosome Location
-
Chr.2R: 21355015 - 21379464 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000395
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080070.4 |
NP_524809.2 |
6 - 7 |
1844 |
65 |
GenBank mRNA |
BT133425.1 |
- |
6 - 7 |
1842 |
65 |
View Full Product Details