Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kn
Entrez Gene ID
45318
Gene Name
knot
Gene Aliases
CG10197, Col, Col/Kn, DmCOE, Dmel\CG10197, Dmel_CG10197, Kn, Knot, coe, col, collier
Chromosome Location
Chr.2R: 14772650 - 14807061 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001319
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103847.2 NP_001097317.1 6 - 7 998 126
NM_001274051.1 NP_001260980.1 5 - 6 667 126
NM_080074.4 NP_524813.2 5 - 6 1119 126
NM_137146.3 NP_610990.2 5 - 6 1119 126
NM_166070.3 NP_725419.2 5 - 6 1119 126
GenBank mRNA BT150395.1 - 5 - 6 1117 126

View Full Product Details