Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aPKC
Entrez Gene ID
47594
Gene Name
atypical protein kinase C
Gene Aliases
CG10261, CG30475, CG42783, DaPKC, Dmel\CG42783, Dmel_CG42783, PKCl, PKCzeta, a-PKC, aPKC zeta, aPKC-zeta, aPKCzeta, aPkc, alphaPKCzeta, apkc, daPKC, dapkc, i, l(2)k06403, pkc-3, psu
Chromosome Location
Chr.2R: 14944458 - 14962969 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261854
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043076.3 NP_001036541.1 9 - 10 2079 72
NM_001043077.3 NP_001036542.1 7 - 8 1718 72
NM_001043078.2 NP_001036543.1 7 - 8 1756 72
NM_001043079.2 NP_001036544.1 7 - 8 1762 72
NM_001043080.3 NP_001036545.1 9 - 10 2015 72
NM_001202010.4 NP_001188939.1 9 - 10 3247 72
NM_001202012.2 NP_001188941.1 8 - 9 1878 72
NM_001274055.1 NP_001260984.1 9 - 10 3268 72
NM_001274056.2 NP_001260985.1 7 - 8 1617 72
NM_001274057.2 NP_001260986.1 7 - 8 1718 72
NM_001274058.2 NP_001260987.1 7 - 8 1718 72
NM_080153.5 NP_524892.2 9 - 10 2201 72
GenBank mRNA BT056247.1 - 9 - 10 2178 72

View Full Product Details