Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- vir
- Entrez Gene ID
-
47869
- Gene Name
- virilizer
- Gene Aliases
-
CG3496, Dmel\CG3496, Dmel_CG3496, Vir
- Chromosome Location
-
Chr.2R: 23360192 - 23366355 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003977
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080161.2 |
NP_524900.1 |
5 - 6 |
5487 |
71 |
View Full Product Details