Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpL4
- Entrez Gene ID
-
49425
- Gene Name
- mitochondrial ribosomal protein L4
- Gene Aliases
-
BG:DS02740.17, CG5818, Dmel\CG5818, Dmel_CG5818, L4, Rpl4, l(2)35Fe, l(2)k14608, l35Fe, mrpL4
- Chromosome Location
-
Chr.2L: 16328059 - 16329132 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001995
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080200.3 |
NP_524939.1 |
1 - 2 |
714 |
88 |
GenBank mRNA |
AY061407.1 |
- |
1 - 2 |
676 |
88 |
View Full Product Details