Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ugt37C1
- Entrez Gene ID
-
53583
- Gene Name
- UDP-glycosyltransferase family 37 member C1
- Gene Aliases
-
AC 004641A, CG8652, DmeUgt37c1, Dmel\CG8652, Dmel_CG8652, EG:EG0003.4, LD15335, UGT37C1, Ugt37c1, ugt37C1
- Chromosome Location
-
Chr.2R: 16843296 - 16845038 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026754
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080268.2 |
NP_525007.1 |
1 - 1 |
1562 |
73 |
GenBank mRNA |
BT132879.2 |
- |
1 - 1 |
1411 |
73 |
View Full Product Details