Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr2a
- Entrez Gene ID
-
31093
- Gene Name
- Gustatory receptor 2a
- Gene Aliases
-
CG18531, Dmel\CG18531, Dmel_CG18531, EG:BACN32G11.1, GR1F.1, GR2B1, GR2a, Gr1F1, Gr2B1
- Chromosome Location
-
Chr.X: 1452430 - 1454177 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0265139
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080299.4 |
NP_525038.3 |
2 - 3 |
1185 |
120 |
GenBank mRNA |
BT122077.2 |
- |
1 - 2 |
983 |
120 |
View Full Product Details