The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272265.1 | NP_001259194.1 | 4 - 5 | 1866 | 54 |
NM_080317.2 | NP_525056.2 | 4 - 5 | 1866 | 54 |