Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rst
Entrez Gene ID
31290
Gene Name
roughest
Gene Aliases
CG4125, CT13684, Dmel\CG4125, Dmel_CG4125, IREE-C, IRREC/RST, Irre-C, IrreC, IrreC-Rst, IrreC-rst, IrreC/Rst, IrrecC, Rst, Rst/IrreC, UB883, irreC, irreC-rst, irrecC-rst, rst-irreC, rst-irrecC, rst/irrecC
Chromosome Location
Chr.X: 2950399 - 2973834 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003285
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297906.1 NP_001284835.1 6 - 7 1575 71
NM_080319.5 NP_525058.2 6 - 7 1575 71
GenBank mRNA BT057980.1 - 6 - 7 1573 71

View Full Product Details