Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptp4E
Entrez Gene ID
31425
Gene Name
Protein tyrosine phosphatase 4E
Gene Aliases
4E, CG6899, CT21187, DPTP4E, Dmel\CG6899, Dmel_CG6899, PTP4E, PtP4E, R-PTP 4E
Chromosome Location
Chr.X: 4939128 - 4955916 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004368
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169198.2 NP_001162669.1 11 - 12 5733 84
NM_001169199.2 NP_001162670.1 11 - 12 5651 84
NM_001169201.2 NP_001162672.1 11 - 12 5652 84
NM_001297946.1 NP_001284875.1 11 - 12 5611 84
NM_080337.3 NP_525076.2 11 - 12 5629 84
GenBank mRNA BT033064.1 - 11 - 12 5624 84

View Full Product Details