Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hexo2
- Entrez Gene ID
-
31808
- Gene Name
- Hexosaminidase 2
- Gene Aliases
-
CG1787, DmHex2, Dmel\CG1787, Dmel_CG1787, HEX 2, HEX2, HEXO2
- Chromosome Location
-
Chr.X: 8709652 - 8712478 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041629
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080342.4 |
NP_525081.1 |
1 - 2 |
303 |
91 |
GenBank mRNA |
AY118361.1 |
- |
1 - 2 |
302 |
91 |
View Full Product Details