Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NaPi-T
Entrez Gene ID
36651
Gene Name
Na[+]-dependent inorganic phosphate cotransporter
Gene Aliases
CG10207, Dmel\CG10207, Dmel_CG10207, Dropo, MFS2, Na/Pi-cotransporter, NapiT
Chromosome Location
Chr.2R: 14842720 - 14845874 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016684
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274052.2 NP_001260981.1 5 - 6 1162 79
NM_080377.3 NP_525116.2 5 - 6 1496 79
GenBank mRNA AY071092.1 - 5 - 6 1159 79

View Full Product Details