Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fal
Entrez Gene ID
43927
Gene Name
falten
Gene Aliases
CG9670, Dmel\CG9670, Dmel_CG9670, Q9VVY4_DROME
Chromosome Location
Chr.3L: 19294564 - 19298830 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028380
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144502.2 NP_001137974.1 2 - 3 503 73
NM_001316453.1 NP_001303382.1 2 - 3 706 73
NM_080528.3 NP_536789.3 5 - 6 1097 73

View Full Product Details