Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
or
Entrez Gene ID
43943
Gene Name
orange
Gene Aliases
AP-3sigma, CG3029, Dmel\CG3029, Dmel_CG3029, Or, aps3, cg3029, sigma3
Chromosome Location
Chr.2R: 23967939 - 23968943 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003008
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080532.4 NP_536793.1 3 - 4 441 78
GenBank mRNA AF228056.1 - 3 - 4 391 78
AY071026.1 - 3 - 4 435 78

View Full Product Details