Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psh
Entrez Gene ID
32832
Gene Name
persephone
Gene Aliases
CG6367, Dmel\CG6367, Dmel_CG6367, PSH, Psh, SP28, c-SP28, cSP28
Chromosome Location
Chr.X: 18484487 - 18486921 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030926
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133069.2 NP_573297.1 3 - 4 564 58
GenBank mRNA AY102662.1 - 3 - 4 564 58

View Full Product Details