Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mec2
Entrez Gene ID
32905
Gene Name
CG7635-PA
Gene Aliases
CG7635, DMec2, Dmel\CG7635, Dmel_CG7635, mec-2
Chromosome Location
Chr.X: 19188035 - 19190193 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030993
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133129.2 NP_573357.1 3 - 4 928 60
GenBank mRNA AY061599.1 - 3 - 4 928 60

View Full Product Details